Få mere at vide
Nitroethane-1,1-d2, CDN
Artikelnummer. 30319978
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319978 5 g
30316049 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319978 Leverandør CDN Leverandørnr. CDND17205G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nitroethane-1,1-D2
CAS 13031-33-9
Molekylær formel C2 D2 H3 N O2
Mængde 5 g
Synonym Ethane-1,1-d2, 1-nitro- (7 CI, 8 CI, 9 CI), 1-Nitroethane-1,1-d2, Nitroethane-d2, Nitroethane-1,1-D2
InChI formel InChI=1 S/C2H5NO2/c1-2-3(4)5/h2H2,1H3/i2D2
SMIL [2 H]C([2 H])(C)[N+](=O)[O-]
IUPAC navn 1,1-dideuterio-1-nitroethane
Molekylvægt (g/mol) 77.079
Formel vægt 77.045 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity