Learn More
Nitrobenzene, TRC
Product Code. 30133864
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133864 10 g 10g
30133476 50 g 50g
30041037 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30133864 Supplier Toronto Research Chemicals Supplier No. TRCN49300010G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 98-95-3
Molecular Formula C6 H5 N O2
Molecular Weight (g/mol) 123.11
Synonym Benzene, nitro-, Nitrobenzene, Essence of Mirbane, Essence of Myrbane, Mirbane oil, NSC 9573, Nitrobenzol, Oil of Mirbane, Oil of Myrbane
IUPAC Name nitrobenzene
SMILES [O-][N+](=O)c1ccccc1
Specifications
Quantity 10 g
InChI Formula InChI=1S/C6H5NO2/c8-7(9)6-4-2-1-3-5-6/h1-5H
Formula Weight 123.032
Percent Purity >95%
Chemical Name or Material Nitrobenzene
Grade HPLC