Få mere at vide
Nitrobenzene Mixture 355 100 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30322812
Change view
Click to view available options
Mængde:
1 ml
Product Code. Mængde
30322812 1 ml
1 options
Product Code. 30322812 Supplier Dr. Ehrenstorfer Supplier No. DREA50000355ME
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Kemisk navn eller materiale 1,2-Dinitrobenzene∣1,3-Dinitrobenzene∣1-Chloro-2-nitrobenzene∣1-Chloro-3-nitrobenzene∣1-Chloro-4-nitrobenzene∣2,5-Dichloronitrobenzene∣3,4-Dichloronitrobenzene∣Nitrobenzene
CAS 528-29-0, 99-65-0, 88-73-3, 121-73-3, 100-00-5, 89-61-2
Analyt- eller komponentnavne 1,2-Dinitrobenzene∣1,3-Dinitrobenzene∣1-Chloro-2-nitrobenzene∣1-Chloro-3-nitrobenzene∣1-Chloro-4-nitrobenzene∣2,5-Dichloronitrobenzene∣3,4-Dichloronitrobenzene∣Nitrobenzene
Forsendelsestilstand Room Temperature
Certificeringer/compliance ISO 17034
Opløsningsmiddel eller Matrix Methanol
Mængde 1 ml
Koncentration eller sammensætning (efter analyt eller komponenter) 1,2-Dinitrobenzene 100 μg/mL∣1,3-Dinitrobenzene 100 μg/mL∣1-Chloro-2-nitrobenzene 100 μg/mL∣1-Chloro-3-nitrobenzene 100 μg/mL∣1-Chloro-4-nitrobenzene 100 μg/mL∣2,5-Dichloronitrobenzene 100 μg/mL∣3,4-Dichloronitrobenzene 100 μg/mL∣Nitrobenzene 100 μg/mL