Få mere at vide
Nitrite Standard: NO2- @ 1000 μg/mL in H2O, VHG™
Artikelnummer. 17999094
Skift visning
Mængde:
100 ml
500 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17999094 500 ml
17918244 100 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17999094 Leverandør VHG Labs Leverandørnr. VHGINO2500
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Nitrite Standard: NO2- @ 1000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nitrite
CAS 14797-65-0
Metoder Ion Chromatography/Wet Chemistry
Analyt- eller komponentnavne Nitrite
Forsendelsestilstand Room Temperature
Molekylær formel (N O2)-
Certificeringer/compliance ISO 17034
Synonym NO2−
SMIL [O-]N=O
IUPAC navn Nitrite
Opløsningsmiddel eller Matrix Water
Molekylvægt (g/mol) 46.01
Mængde 500 ml
Formel vægt 46.01 g/mol