Få mere at vide
Nitrilotriacetic Acid-d9, TRC
Artikelnummer. 30477012
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30477012 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30477012 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49017625MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nitrilotriacetic Acid-d9
CAS 807630-34-8
Molekylær formel C6 D9 N O6
Mængde 25 mg
Synonym Glycine-1,2,2-d3','N,N-bis[(carboxy-d)methyl-d2]- (9CI), N,N-Bis[(carboxy-d)methyl-d2]glycine-1,2,2-d3, Nitrilotriacetic Acid-d9, Nitrilotriacetic Acid-D9, NTA-d9, Deuterio 2-[bis(1,1-dideuterio-2-deuteriooxy-2-oxo-ethyl)amino]-2,2-dideuterio-acetate
InChI formel InChI=1S/C6H9NO6/c8-4(9)1-7(2-5(10)11)3-6(12)13/h1-3H2,(H,8,9)(H,10,11)(H,12,13)/i1D2,2D2,3D2/hD3
SMIL [2H]OC(=O)C([2H])([2H])N(C([2H])([2H])C(=O)O[2H])C([2H])([2H])C(=O)O[2H]
IUPAC navn deuterio 2-[bis(1,1-dideuterio-2-deuteriooxy-2-oxoethyl)amino]-2,2-dideuterioacetate
Molekylvægt (g/mol) 200.1942
Formel vægt 200.0995