missing translation for 'onlineSavingsMsg'
Learn More

Nitrilotriacetic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-7081795
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
139-13-9
C6H9NO6
191.14
MFCD00004287
MGFYIUFZLHCRTH-UHFFFAOYSA-N
nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
8758
CHEBI:44557
2-[bis(carboxymethyl)amino]acetic acid
OC(=O)CN(CC(O)=O)CC(O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
139-13-9
C6H9NO6
191.14
MFCD00004287
MGFYIUFZLHCRTH-UHFFFAOYSA-N
nitrilotriacetic acid, 2,2',2-nitrilotriacetic acid, triglycollamic acid, aminotriacetic acid, complexon i, trilon a, n,n-bis carboxymethyl glycine, nitrilotriacetate, komplexon i, titriplex i
8758
CHEBI:44557
2-[bis(carboxymethyl)amino]acetic acid
OC(=O)CN(CC(O)=O)CC(O)=O

It is employed as an excellent chelating agent. It is also used as a complexing agent, metal masking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an excellent chelating agent. It is also used as a complexing agent, metal masking agent.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point 246°C (decomposition)
Odor Odorless
Linear Formula N(CH2CO2H)3
Quantity 100 g
Beilstein 1710776
Merck Index 14,6579
Solubility Information Slightly soluble in water.
Formula Weight 191.14
Percent Purity 99%
Chemical Name or Material Nitrilotriacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'