Få mere at vide
Nitrefazole, TRC
Artikelnummer. 30474911
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30474911 2.5 g 2.5g
30474079 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30474911 Leverandør Toronto Research Chemicals Leverandørnr. TRCN4901252.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 21721-92-6
Molekylær formel C10H8N4O4
Molekylvægt (g/mol) 248.19
Synonym 2-Methyl-4-nitro-1-(p-nitrophenyl)-Imidazole, 1-(4-Nitrophenyl)-2-methyl-4-nitroimidazole, 2-Methyl-4-nitro-1-(4-nitrophenyl)-1H-imidazole, Altimol, EMD 15700, Nitrefazole
IUPAC navn 2-methyl-4-nitro-1-(4-nitrophenyl)imidazole
SMIL Cc1nc(cn1c2ccc(cc2)[N+](=O)[O-])[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale Nitrefazole
Mængde 2.5 g
InChI formel InChI=1S/C10H8N4O4/c1-7-11-10(14(17)18)6-12(7)8-2-4-9(5-3-8)13(15)16/h2-6H,1H3
Formel vægt 248.0546