missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nitinol compression coil spring, wire diameter 1.0mm (0.039in), mean coil diameter 7.0mm (0.28in), Alloy M

Brand:  Alfa Aesar™ 045951.KJ

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15484025

  • 378.86 DKK / Pack of 5


Estimated Shipment
23-10-2020
Add to basket

Specifications

Specifications

Nitinol compression coil spring, Alloy M
Wire diameter 1.0mm (0.039 in.), mean coil diameter 7.0mm (0.27 in.), mean coil diameter 7.0mm (0.28 in.)
Odorless
5 pc.
MFCD02091734
Af approx. 41°C
Videos
SDS Documents