Få mere at vide
NITD-349, MedChemExpress
Artikelnummer. 16497027
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16497027 5 mg -
16485226 100 mg 100mg
16480637 50 mg -
16404318 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16497027 Leverandør Medchem Express Leverandørnr. HY109588
MedChemExpress NITD-349 is an MmpL3 inhibitor that shows highly potent anti-mycobacterial activity with MIC50 of 23 nM against virulent Mycobacterium tuberculosis H37Rv.
NITD-349 is an MmpL3 inhibitor that shows highly potent anti-mycobacterial activity with MIC50 of 23 nM against virulent Mycobacterium tuberculosis H37Rv.
Tekniske data
Kemisk navn eller materiale NITD-349
Fysisk form Solid
CAS 1473450-62-2
Farve White
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C17H20F2N2O
Til brug med (applikation) COVID-19-immunoregulation
Opløselighedsinformation DMSO : ≥ 310 mg/mL (1011.91 mM)
SMIL O=C(C(N1)=CC2=C1C=C(F)C=C2F)NC3CCC(C)(C)CC3
Molekylvægt (g/mol) 306.35
Formel vægt 306.35
Procent renhed 98.84%