Få mere at vide
Nitazoxanide, TRC
Artikelnummer. 30134177
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134177 25 mg
30138653 100 mg
17963995 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134177 Leverandør Toronto Research Chemicals Leverandørnr. TRCN49010025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 55981-09-4
Molekylær formel C12 H9 N3 O5 S
Molekylvægt (g/mol) 307.28
Synonym Benzamide, 2-(acetyloxy)-N-(5-nitro-2-thiazolyl)-, Alinia, BRN 1225475, Cryptaz, DRG 0242, Daxon, Dexidex, Kidonax, Mitafar, N-(5-Nitro-2-thiazolyl)salicylamide acetate (ester), NSC 697855, NTX, NTZ, Nitacure, Nitacure 500, Nitarid, Nitazode, Nitazox 500, Nitazoxanida, Nitazoxanide, Nixide, Nixoran, Nizonide, PH 5776, Pacovanton, ParaMix, Xerovirin-C
IUPAC navn [2-[(5-nitro-1,3-thiazol-2-yl)carbamoyl]phenyl] acetate
SMIL CC(=O)Oc1ccccc1C(=O)Nc2ncc(s2)[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale Nitazoxanide
Mængde 25 mg
InChI formel InChI=1S/C12H9N3O5S/c1-7(16)20-9-5-3-2-4-8(9)11(17)14-12-13-6-10(21-12)15(18)19/h2-6H,1H3,(H,13,14,17)
Formel vægt 307.0263