Få mere at vide
Nintedanib Carboxylic Acid, TRC
Artikelnummer. 30010505
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010505 50 mg
30010742 25 mg
30011432 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010505 Leverandør Toronto Research Chemicals Leverandørnr. TRCN47828550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 894783-71-2
Molekylær formel C30 H31 N5 O4
Molekylvægt (g/mol) 525.6
Synonym (3Z)-2,3-Dihydro-3-[[[4-[methyl[(4-methyl-1-piperazinyl)acetyl]amino]phenyl]amino]phenylmethylene]-2-oxo-1H-Indole-6-carboxylic Acid, (3Z)-2,3-Dihydro-3-[[[4-[methyl[2-(4-methyl-1-piperazinyl)acetyl]amino]phenyl]amino]phenylmethylene]-2-oxo-1H-indole-6-carboxylic Acid, BIBF 1202
IUPAC navn (3Z)-3-[[4-[methyl-[2-(4-methylpiperazin-1-yl)acetyl]amino]anilino]-phenyl-methylene]-2-oxo-indoline-6-carboxylic acid
SMIL CN(C(=O)CN1CCN(C)CC1)c2ccc(N\C(=C\3/C(=O)Nc4cc(ccc34)C(=O)O)\c5ccccc5)cc2
Tekniske data
Kemisk navn eller materiale Nintedanib Carboxylic Acid
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C30H31N5O4/c1-33-14-16-35(17-15-33)19-26(36)34(2)23-11-9-22(10-12-23)31-28(20-6-4-3-5-7-20)27-24-13-8-21(30(38)39)18-25(24)32-29(27)37/h3-13,18,31H,14-17,19H2,1-2H3,(H,32,37)(H,38,39)/b28-27-
Formel vægt 525.2376
Procent renhed >95%