Learn More
Thermo Scientific Chemicals Ninhydrin, spectrophotometric grade
Catalog Number p-172476
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
485-47-2
C9H6O4
178.143
MFCD00003791
FEMOMIGRRWSMCU-UHFFFAOYSA-N
1, 2, 3-Indantrione, 1, 2, 3-Triketohydrindene, 2, 2-Dihydroxy-1H-indene-1, 3(2H)-dione, 1H-Indene-1, 2, 3-trione
10236
CHEBI:86374
2,2-dihydroxyindene-1,3-dione
C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
485-47-2
C9H6O4
178.143
MFCD00003791
FEMOMIGRRWSMCU-UHFFFAOYSA-N
1, 2, 3-Indantrione, 1, 2, 3-Triketohydrindene, 2, 2-Dihydroxy-1H-indene-1, 3(2H)-dione, 1H-Indene-1, 2, 3-trione
10236
CHEBI:86374
2,2-dihydroxyindene-1,3-dione
C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 250.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, slightly soluble in ether and chloroform
Infrared Spectrum Authentic
Formula Weight 178.14
Grade Spectrophotometric
Packaging Glass bottle
Suitability (for amino acid analysis): passes
Density 0.81
Color Yellow
pH 4.6 to 5.6 (1% aq. soln.)
Quantity 25 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Ninhydrin
Show More Show Less
compliance-icons
Product Identifier
  • Ninhydrin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.