Få mere at vide
Nimodipine-d7, TRC
Artikelnummer. 30472358
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30472358 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472358 Leverandør Toronto Research Chemicals Leverandørnr. TRCN4782025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nimodipine-d7
CAS 1246815-36-0
Renhedsgrad noter HPLC
Molekylær formel C21H19D7N2O7
Mængde 5 mg
Synonym 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 3-(2-Methoxyethyl) 5-(1-Methylethyl-d7) Ester, (+/-)-Nimodipine-d7, Admon-d7, BAY-e 9736-d7, Nimodipine AP-d7, Nimotop-d7, Periplum-d7
InChI formel InChI=1S/C21H26N2O7/c1-12(2)30-21(25)18-14(4)22-13(3)17(20(24)29-10-9-28-5)19(18)15-7-6-8-16(11-15)23(26)27/h6-8,11-12,19,22H,9-10H2,1-5H3/i1D3,2D3,12D
SMIL O=C(OCCOC)C1=C(C)NC(C)=C(C(OC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)C1C2=CC=CC([N+]([O-])=O)=C2
Molekylvægt (g/mol) 425.48
Formel vægt 425.2179
Procent renhed >95