Få mere at vide
Nimesulide-d5, TRC
Artikelnummer. 30472703
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30472703 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472703 Leverandør Toronto Research Chemicals Leverandørnr. TRCN47750225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nimesulide-d5
CAS 1330180-22-7
Renhedsgrad noter HPLC
Molekylær formel C13H7D5N2O5S
Mængde 25 mg
Synonym N-(4-Nitro-2-(phenoxy-d5)phenyl)methanesulfonamide, 4-Nitro-2-(phenoxy-d5) methanesulfonamide-d5, 4'-Nitro-2'-(phenoxy-d5)methanesulfonanilide, Aulin-d5, Flogovital-d5, Lizepat-d5, Mesulid-d5, Nimed-d5, Nimepast-d5, Nimsulid-d5, Nimulid-d5, Nise*Gel-d5, Nisulid-d5, Nizer-d5, Orthobid-d5, R 805-d5, Sulide-d5, Sulidene-d5
InChI formel InChI=1S/C13H12N2O5S/c1-21(18,19)14-12-8-7-10(15(16)17)9-13(12)20-11-5-3-2-4-6-11/h2-9,14H,1H3/i2D,3D,4D,5D,6D
SMIL CS(NC1=CC=C([N+]([O-])=O)C=C1OC2=C([2H])C([2H])=C([2H])C([2H])=C2[2H])(=O)=O
IUPAC navn N-(4-nitro-2-(phenoxy-d5)phenyl)methanesulfonamide
Molekylvægt (g/mol) 313.34
Formel vægt 313.08
Procent renhed >95