missing translation for 'onlineSavingsMsg'
Få mere at vide

Nifurstyrenic acid sodium 100 μg/mL in Acetonitrile:Water, Dr. Ehrenstorfer

Artikelnummer. 30316188
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30316188 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30316188 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA15525100WL100

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Nifurstyrenic Acid Sodium
CAS 54992-23-3
Analyt- eller komponentnavne Nifurstyrenic Acid Sodium
Forsendelsestilstand Room Temperature
Molekylær formel C13 H8 N O5 . Na
Koncentration 100 μg/mL
Certificeringer/compliance ISO 17034
Synonym Benzoic acid, 4-[2-(5-nitro-2-furanyl)ethenyl]-, sodium salt (1:1), Benzoic acid, 4-[2-(5-nitro-2-furanyl)ethenyl]-, sodium salt (9CI), Nifurstyrenate sodium, Sodium nifurstyrenate
InChI formel InChI=1S/C13H9NO5.Na/c15-13(16)10-4-1-9(2-5-10)3-6-11-7-8-12(19-11)14(17)18;/h1-8H,(H,15,16);/q;+1/p-1/b6-3+;
SMIL [Na+].[O-]C(=O)c1ccc(\C=C\c2oc(cc2)[N+](=O)[O-])cc1
IUPAC navn sodium;4-[(E)-2-(5-nitrofuran-2-yl)ethenyl]benzoate
Opløsningsmiddel eller Matrix Acetonitrile/Water
Molekylvægt (g/mol) 281.2
Mængde 1 ml
Formel vægt 281.03 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.