Få mere at vide
Niflumic Acid EP Impurity F, Mikromol™
Beskrivelse
Niflumic Acid EP Impurity F
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl 2-[(3-(Trifluoromethyl)phenyl)amino]pyridine-3-carboxylate (Niflumic Acid Methyl Ester) |
| CAS | 59361-45-4 |
| Analyt- eller komponentnavne | Methyl 2-[(3-(Trifluoromethyl)phenyl)amino]pyridine-3-carboxylate (Niflumic Acid Methyl Ester) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C14 H11 F3 N2 O2 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Methyl 2-[[3-(Trifluoromethyl)phenyl]amino]pyridine-3-carboxylate (Niflumic Acid Methyl Ester), Methyl 2-[[3-(Trifluoromethyl)phenyl]amino]pyridine-3-carboxylate, Niflumic Acid Imp. F (EP), Niflumic Acid Methyl Ester, Niflumic Acid Impurity F |
| InChI formel | InChI=1S/C14H11F3N2O2/c1-21-13(20)11-6-3-7-18-12(11)19-10-5-2-4-9(8-10)14(15,16)17/h2-8H,1H3,(H,18,19) |
| SMIL | COC(=O)c1cccnc1Nc2cccc(c2)C(F)(F)F |
| IUPAC navn | methyl 2-[3-(trifluoromethyl)anilino]pyridine-3-carboxylate |
| Vis mere |