Få mere at vide
Niflumic Acid-d5 (Major), TRC
Artikelnummer. 30476237
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30476237 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476237 Leverandør Toronto Research Chemicals Leverandørnr. TRCN4575021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Niflumic Acid-d5
CAS 1794811-58-7
Molekylær formel C13H4D5F3N2O2
Mængde 1 mg
Synonym 2-[[3-(Trifluoromethyl)phenyl]amino]-3-pyridinecarboxylic Acid-d5, 2-[3-(Trifluoromethyl)anilino]nicotinic Acid-d5, Actol-d5, Donalgin-d5, Flogovital-d5, Forenol-d5, Landruma-d5, Nifluminic Acid-d5, Nifluril-d5, UP 83-d5
InChI formel InChI=1S/C13H9F3N2O2/c14-13(15,16)8-3-1-4-9(7-8)18-11-10(12(19)20)5-2-6-17-11/h1-7H,(H,17,18)(H,19,20)/i1D,2D,4D,5D,6D
SMIL [2H]C1=C([2H])C([2H])=C(C(O)=O)C(NC2=C([2H])C([2H])=CC(C(F)(F)F)=C2)=N1
IUPAC navn 4,5,6-trideuterio-2-[2,3-dideuterio-5-(trifluoromethyl)anilino]pyridine-3-carboxylic acid
Molekylvægt (g/mol) 287.25
Formel vægt 287.093