Få mere at vide
Niflumic acid, 99+%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J60489.06
| Mængde | 5g |
|---|
Se flere versioner af dette produkt
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used as a selective Cox-2 inhibitor and GPR35 activator. Also activates human TRPA1 inducibly expressed in HEK293 cells; displays agonist activity at GPR35.
Solubility
Soluble in ethanol (∼50 mg/ml), acetone (50 mg/ml, Clear to Slightly Hazy, Yellow), methanol (∼50 mg/ml), DMSO (56 mg/ml at 25°C), and acetonitrile (∼50 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Hygroscopic. Keep away fom strong oxidizing agents.
Kemiske identifikatorer
| 4394-00-7 | |
| 282.22 | |
| JZFPYUNJRRFVQU-UHFFFAOYSA-N | |
| 4488 | |
| 2-{[3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid |
| C13H9F3N2O2 | |
| MFCD00010569 | |
| niflumic acid, nifluril, landruma, forenol, actol, acido niflumico, nifluminic acid, acide niflumique, acidum niflumicum, acide niflumique inn-french | |
| CHEBI:34888 | |
| OC(=O)C1=CC=CN=C1NC1=CC=CC(=C1)C(F)(F)F |
Tekniske data
| Niflumic acid | |
| C13H9F3N2O2 | |
| 5g | |
| Light sensitive | |
| niflumic acid, nifluril, landruma, forenol, actol, acido niflumico, nifluminic acid, acide niflumique, acidum niflumicum, acide niflumique inn-french | |
| JZFPYUNJRRFVQU-UHFFFAOYSA-N | |
| 2-{[3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid | |
| 4488 | |
| 282.22 |
| 4394-00-7 | |
| MFCD00010569 | |
| UN2811 | |
| 14,6531 | |
| Soluble in ethanol (∽50mg/ml),acetone (50mg/ml,Clear to Slightly Hazy,Yellow),methanol (∽50mg/ml),DMSO (56mg/ml at 25°C),and acetonitrile (∽50mg/ml). | |
| OC(=O)C1=CC=CN=C1NC1=CC=CC(=C1)C(F)(F)F | |
| 282.22 | |
| CHEBI:34888 | |
| ≥99% |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.