Få mere at vide
Nifenazone, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Nifenazone |
| CAS | 2139-47-1 |
| Molekylær formel | C17 H16 N4 O2 |
| Mængde | 1 g |
| Synonym | 3-Pyridinecarboxamide','N-(2,3-dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)-, Nicotinamide','N-antipyrinyl- (6CI,7CI,8CI), 4-(Nicotinoylamino)antipyrine, 4-Nicotinamido-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Dolongan, N-Antipyrinylnicotinamide, N-Nicotinoylaminoantipyrine, Niapyrin, Nicazane, Nicodon, Nicodone, Nicofesone, Nicophesone, Nicophezon, Nicopyron, Nicoreumal, Nicoron, Nicotazone, Nicotinic acid 4-antipyrilamide, Nicotinoylantipyrine, Nicozone, Nifenazon, Nifenazone, Nikodon, Nikofezon, Niprazine, Phenicazone, Reupiron, Thylin |
| InChI formel | InChI=1S/C17H16N4O2/c1-12-15(19-16(22)13-7-6-10-18-11-13)17(23)21(20(12)2)14-8-4-3-5-9-14/h3-11H,1-2H3,(H,19,22) |
| SMIL | CN1N(C(=O)C(=C1C)NC(=O)c2cccnc2)c3ccccc3 |
| IUPAC navn | N-(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)pyridine-3-carboxamide |
| Molekylvægt (g/mol) | 308.33 |
| Formel vægt | 308.1273 |