Få mere at vide
Nifedipine Monoamide, TRC
Artikelnummer. 30479590
Skift visning
Mængde:
25 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479590 25 mg
30473457 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30479590 Leverandør Toronto Research Chemicals Leverandørnr. TRCN45705025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nifedipine Monoamide
CAS 114709-68-1
Molekylær formel C16 H17 N3 O5
Mængde 25 mg
Synonym 3-Pyridinecarboxylic acid','5-(aminocarbonyl)-1,4-dihydro-2,6-dimethyl-4-(2-nitrophenyl)-','methyl ester, Methyl 5-Carbamoyl-2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3-carboxylate
InChI formel InChI=1S/C16H17N3O5/c1-8-12(15(17)20)14(13(9(2)18-8)16(21)24-3)10-6-4-5-7-11(10)19(22)23/h4-7,14,18H,1-3H3,(H2,17,20)
SMIL COC(=O)C1=C(C)NC(=C(C1c2ccccc2[N+](=O)[O-])C(=O)N)C
IUPAC navn methyl 5-carbamoyl-2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3-carboxylate
Molekylvægt (g/mol) 331.32
Formel vægt 331.1168