Få mere at vide
Nicotinuric Acid-d4, TRC
Artikelnummer. 30468327
Skift visning
Mængde:
2.5 mg
10 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30468327 10 mg -
30464745 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30468327 Leverandør Toronto Research Chemicals Leverandørnr. TRCN43350210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1216737-36-8
Molekylær formel C8 2H4 H4 N2 O3
Molekylvægt (g/mol) 184.19
Synonym 2-[(2,4,5,6-Tetradeuteriopyridine-3-carbonyl)amino]acetic acid, Nicotinuric-d4 Acid (pyridyl-d4), Nicotinuric Acid-d4, Nicotinuric Acid-D4 (pyridyl-D4), (Nicotinoyl-d4-amino)acetic acid, 2-(Nicotinylamino-2,4,5,6-d4)acetic acid, Nicotinoyl-d4-glycine
IUPAC navn 2-[(2,4,5,6-tetradeuteriopyridine-3-carbonyl)amino]acetic acid
SMIL [2H]c1nc([2H])c(C(=O)NCC(=O)O)c([2H])c1[2H]
Tekniske data
Kemisk navn eller materiale Nicotinuric Acid-d4
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C8H8N2O3/c11-7(12)5-10-8(13)6-2-1-3-9-4-6/h1-4H,5H2,(H,10,13)(H,11,12)/i1D,2D,3D,4D
Formel vægt 184.0786
Procent renhed >95%