Få mere at vide
Nicotinoyl-gamma-aminobutyric Acid, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Nicotinoyl-gamma-aminobutyric Acid |
| CAS | 34562-97-5 |
| Molekylær formel | C10H12N2O3 |
| Mængde | 500 mg |
| Synonym | 4-Nicotinamidobutyric Acid, 4-[(3-Pyridinylcarbonyl)amino]butanoic Acid, N-Nicotinoyl-γ-aminobutyric Acid, Nicotinoyl-GABA |
| InChI formel | InChI=1S/C10H12N2O3/c13-9(14)4-2-6-12-10(15)8-3-1-5-11-7-8/h1,3,5,7H,2,4,6H2,(H,12,15)(H,13,14) |
| SMIL | O=C(O)CCCNC(C1=CN=CC=C1)=O |
| Molekylvægt (g/mol) | 208.21 |