Få mere at vide
Nicotinic-d4 Acid, CDN
Artikelnummer. 30315356
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315356 0.25 g
30313652 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30315356 Leverandør CDN Leverandørnr. CDND43680.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nicotinic-d4 Acid
CAS 66148-15-0
Molekylær formel C6 2H4 H N O2
Mængde 0.25 g
Synonym Nicotinic Acid-D4, 3-Pyridine-2,4,5,6-d4-carboxylic acid
InChI formel InChI=1 S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4 H,(H,8,9)/i1D,2 D,3 D,4 D
SMIL [2 H]c1nc([2 H])c(C(=O)O)c([2 H])c1[2 H]
IUPAC navn 2,4,5,6-tetradeuteriopyridine-3-carboxylic acid
Molekylvægt (g/mol) 127.13
Formel vægt 127.0571 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity