missing translation for 'onlineSavingsMsg'
Få mere at vide

Nicotinic Acid-d4, TRC

Artikelnummer. 30467681
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30467681 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30467681 Leverandør Toronto Research Chemicals Leverandørnr. TRCN429253100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Nicotinic-d4 Acid
CAS 66148-15-0
Renhedsgrad noter HPLC
Molekylær formel C6 2H4 H N O2
Mængde 100 mg
Synonym Nicotinic Acid-D4, 3-Pyridine-2,4,5,6-d4-carboxylic acid
InChI formel InChI=1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)/i1D,2D,3D,4D
SMIL [2H]c1nc([2H])c(C(=O)O)c([2H])c1[2H]
IUPAC navn 2,4,5,6-tetradeuteriopyridine-3-carboxylic acid
Molekylvægt (g/mol) 127.13
Formel vægt 127.0571
Procent renhed >95
Vis mere Vis mindre
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.