Få mere at vide
Nicotinic Acid-d4, TRC
Artikelnummer. 30472977
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30472977 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472977 Leverandør Toronto Research Chemicals Leverandørnr. TRCN4292532.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nicotinic-d4 Acid
CAS 66148-15-0
Renhedsgrad noter HPLC
Molekylær formel C6 2H4 H N O2
Mængde 2.5 mg
Synonym Nicotinic Acid-D4, 3-Pyridine-2,4,5,6-d4-carboxylic acid
InChI formel InChI=1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)/i1D,2D,3D,4D
SMIL [2H]c1nc([2H])c(C(=O)O)c([2H])c1[2H]
IUPAC navn 2,4,5,6-tetradeuteriopyridine-3-carboxylic acid
Molekylvægt (g/mol) 127.13
Formel vægt 127.0571
Procent renhed >95