Få mere at vide
Nicotinic Acid-d3 (major), TRC
Artikelnummer. 30471774
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30471774 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30471774 Leverandør Toronto Research Chemicals Leverandørnr. TRCN42924825MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nicotinic Acid-d3 (major)
CAS 861405-75-6
Molekylær formel C6H2D3NO2
Mængde 25 mg
Synonym Nicotinic Acid D3
InChI formel InChI=1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)/i1D,3D,4D
SMIL [2H]c1cc(C(=O)O)c([2H])nc1[2H]
Molekylvægt (g/mol) 126.13
Formel vægt 126.0509