Få mere at vide
Nicotinic Acid-13C6,d4 N-Hydroxysuccinimide Ester, TRC
Artikelnummer. 30467897
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30467897 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30467897 Leverandør Toronto Research Chemicals Leverandørnr. TRCN4292575MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nicotinic Acid-13C6','d4 N-Hydroxysuccinimide Ester
CAS 1209457-55-5
Molekylær formel 13C6 C4 D4 H4 N2 O4
Mængde 5 mg
Synonym Nicotinic Acid 13C6,D4 N-Hydroxysuccinimide Ester
InChI formel InChI=1S/C10H8N2O4/c13-8-3-4-9(14)12(8)16-10(15)7-2-1-5-11-6-7/h1-2,5-6H,3-4H2/i1+1D,2+1D,5+1D,6+1D,7+1,10+1
SMIL [2H][13c]1n[13c]([2H])[13c]([13C](=O)ON2C(=O)CCC2=O)[13c]([2H])[13c]1[2H]
IUPAC navn (2,5-dioxopyrrolidin-1-yl) 2,4,5,6-tetradeuterio(2,3,4,5,6-13C5)pyridine-3-(13C)carboxylate
Molekylvægt (g/mol) 230.162
Formel vægt 230.094