Få mere at vide
(±)-Nicotine-d3 (N-methyl-d3), CDN
Artikelnummer. 30321553
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321553 0.1 g 0.1g
30318875 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321553 Leverandør CDN Leverandørnr. CDND35190.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (+/-)-Nicotine-d3
CAS 69980-24-1
Molekylær formel C10 2H3 H11 N2
Mængde 0.1 g
Synonym 3-[1-(Methyl-d3)-2-pyrrolidinyl]pyridine, (±)-3-[1-(Methyl-d3)-2-pyrrolidinyl]pyridine, (±)-Nicotine-N-(methyl-d3), dl-Nicotine-N-(methyl-d3)
InChI formel InChI=1 S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10 H,3,5,7H2,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])N1CCCC1c2cccnc2
IUPAC navn 3-[1-(trideuteriomethyl)pyrrolidin-2-yl]pyridine
Molekylvægt (g/mol) 165.25
Formel vægt 165.1345 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity