Få mere at vide
Nicotinamide-d4, TRC
Artikelnummer. 30471166
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30471166 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30471166 Leverandør Toronto Research Chemicals Leverandørnr. TRCN40775225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nicotinamide-2,4,5,6-D4
CAS 347841-88-7
Renhedsgrad noter HPLC
Molekylær formel C6 2H4 H2 N2 O
Mængde 25 mg
Synonym Nicotinamide D4 (2,4,5,6 D4), 3-Pyridine-2,4,5,6-d4-carboxamide, Nicotinamide-d4, Nicotinamide-2,4,5,6-D4, 3-(Aminocarbonyl)pyridine-2,4,5,6-d4
InChI formel InChI=1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)/i1D,2D,3D,4D
SMIL [2H]c1nc([2H])c(C(=O)N)c([2H])c1[2H]
IUPAC navn 2,4,5,6-tetradeuteriopyridine-3-carboxamide
Molekylvægt (g/mol) 126.15
Formel vægt 126.0731
Procent renhed >95