Få mere at vide
Nicotinamide-d4 N-Oxide (d4 Major), TRC
Artikelnummer. 30469143
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30469143 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469143 Leverandør Toronto Research Chemicals Leverandørnr. TRCN40776250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nicotinamide-d4 N-Oxide (d4 Major)
CAS 1246817-64-0
Molekylær formel C6H2D4N2O2
Mængde 50 mg
Synonym 2,4,5,6-Tetradeuterio-1-oxidopyridin-1-ium-3-carboxamide, 3-Pyridinecarboxamide-2,4,5,6-d4 1-oxide, Nicotinamide-2,4,5,6-d4 1-oxide, Niacinamide-d4 N-oxide, Nicotinamide-d4 N-oxide, Nicotinamide N-oxide-d4, Nicotinamide N-oxide D4 (pyridine-2,4,5,6-D4), Pyridine-2,4,5,6-d4-3-carboxamide 1-oxide
InChI formel InChI=1S/C6H6N2O2/c7-6(9)5-2-1-3-8(10)4-5/h1-4H,(H2,7,9)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(C(=O)N)c([2H])[n+]([O-])c1[2H]
Molekylvægt (g/mol) 142.15
Formel vægt 142.068