Få mere at vide
Nicotelline, TRC
Artikelnummer. 30133823
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133823 50 mg
30004364 5 mg
30010675 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133823 Leverandør Toronto Research Chemicals Leverandørnr. TRCN40120050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 494-04-2
Molekylær formel C15 H11 N3
Molekylvægt (g/mol) 233.27
IUPAC navn 2,4-dipyridin-3-ylpyridine
SMIL c1cncc(c1)c2ccnc(c2)c3cccnc3
Tekniske data
Kemisk navn eller materiale Nicotelline
Mængde 50 mg
InChI formel InChI=1S/C15H11N3/c1-3-13(10-16-6-1)12-5-8-18-15(9-12)14-4-2-7-17-11-14/h1-11H
Formel vægt 233.0953