Få mere at vide
Nicosulfuron-d6, TRC
Artikelnummer. 30473501
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30473501 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473501 Leverandør Toronto Research Chemicals Leverandørnr. TRCN39950210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Nicosulfuron-d6
CAS 1189419-41-7
Renhedsgrad noter HPLC
Molekylær formel C15 2H6 H12 N6 O6 S
Mængde 10 mg
Synonym Nicosulfuron D6 (Dimethoxy D6), 2-[[4,6-Bis(trideuteriomethoxy)pyrimidin-2-yl]carbamoylsulfamoyl]-N,N-dimethylpyridine-3-carboxamide, Nicosulfuron D6 (Dimethoxy D6), Nicosulfuron D6 (dimethoxy D6), 2-[[4,6-Bis(trideuteriomethoxy)pyrimidin-2-yl]carbamoylsulfamoyl]-N,N-dimethylpyridine-3-carboxamide
InChI formel InChI=1S/C15H18N6O6S/c1-21(2)13(22)9-6-5-7-16-12(9)28(24,25)20-15(23)19-14-17-10(26-3)8-11(18-14)27-4/h5-8H,1-4H3,(H2,17,18,19,20,23)/i3D3,4D3
SMIL [2H]C([2H])([2H])Oc1cc(OC([2H])([2H])[2H])nc(NC(=O)NS(=O)(=O)c2ncccc2C(=O)N(C)C)n1
IUPAC navn 2-[[4,6-bis(trideuteriomethoxy)pyrimidin-2-yl]carbamoylsulfamoyl]-N,N-dimethylpyridine-3-carboxamide
Molekylvægt (g/mol) 416.44
Formel vægt 416.1385
Procent renhed >95