Learn More
Nickel Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
442.00 DKK - 733.00 DKK
Chemical Identifiers
CAS | 7440-02-0, 7697-37-2, 7732-18-5 |
---|---|
Molecular Formula | Ni |
Molecular Weight (g/mol) | 58.69 |
MDL Number | MFCD00011137 MFCD06798735 |
InChI Key | PXHVJJICTQNCMI-UHFFFAOYSA-N |
PubChem CID | 935 |
ChEBI | CHEBI:28112 |
IUPAC Name | nickel |
SMILES | [Ni] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
16381166
|
SPEX Certiprep
CLNI2-2M |
30 mL | Triple-leached LDPE bottle |
442.00 DKK
30mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10321224
|
SPEX Certiprep
CLNI2-2Y |
125 mL | Triple-leached LDPE bottle |
733.00 DKK
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ni | 1000ÎĽg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7440-02-0, 7697-37-2, 7732-18-5 | |
58.69 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
CHEBI:28112 | |
[Ni] |
Ni | |
MFCD00011137 MFCD06798735 | |
935 | |
nickel |
Specifications
7440-02-0, 7697-37-2, 7732-18-5 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Ni | |
ICP-MS | |
Standard Solution | |
ICP/ICP-MS Standard | |
nickel | |
58.69 | |
CHEBI:28112 | |
Nickel Standard |
0.1, 2.0, 97.9 | |
0.1, 2.0, 97.9 | |
MFCD00011137 MFCD06798735 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
[Ni] | |
2% HNO3 | |
935 | |
ICP/ICP-MS |
Safety and Handling
Causes severe skin burns and eye damage
missing translation for 'dotInformation' : Nonhazardous
Recommended Storage : Ambient
- 1000
Signal Word
- Danger||Warning
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P364-And wash it before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitric acid, nickel
Product intended for laboratory use only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.