missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Nickel, Ni/Cr/Mo/Nb, Alloy 625, N06625, ARMI∣MBH
Description
Nickel, Ni/Cr/Mo/Nb, Alloy 625, N06625
Specifications
Specifications
| Kemisk navn eller materiale | Aluminium, Boron, Carbon, Chromium, Cobalt, Copper, Iron, Magnesium, Manganese, Molybdenum, Nickel, Niobium, Nitrogen, Oxygen, Phosphorus, Silicon, Sulfur, Tantalum, Tin, Titanium, Tungsten, Vanadium, Zirconium |
| Mængde | 1 Ea. |
| Diameter (metrisk) Udvendig | 38 mm |
| Form | Disc |
| Tykkelse (metrisk) | 19 mm |
| Koncentration eller sammensætning (efter analyt eller komponenter) | Tin: 7 ppm, Oxygen: 6 ppm, Zirconium: 6 ppm, Sulfur: 0.0004%, Magnesium: 0.0008%, Boron: 0.0019%, Tantalum: 0.004%, Phosphorus: 0.007%, Nitrogen: 0.01%, Vanadium: 0.016%, Carbon: 0.031%, Copper: 0.076%, Silicon: 0.076%, Manganese: 0.08%, Tungsten: 0.088%, Cobalt: 0.116%, Titanium: 0.296%, Aluminium: 0.303%, Niobium: 3.5%, Iron: 3.98%, Molybdenum: 8.74%, Chromium: 22.27%, Nickel: 60.4% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?