missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Chromium lump, 99.9% (metals basis), Thermo Scientific Chemicals
621.03 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
For vacuum deposition. hey are also used in some dental restorations. Used as Heating Elements, Thermocouples, Wear Resistant Alloys. Used as intermediate and research. Nichrome wire is commonly used in ceramics and Pyrotechnics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Avg. Mol. Wt. or Mol. Wt. Range | Ni:Cr; 80:20 wt% |
Quantity | 25 g |
Solubility Information | Insoluble in water. |
Percent Purity | 99.9% |
Odor | Odorless |
Particle Size | 3 to 12 mm |
Form | Sintered Lump |
Assay | Metals basis |
Chemical Name or Material | Nickel Chromium lump |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'