missing translation for 'onlineSavingsMsg'
Få mere at vide

Nicergoline, TRC

Artikelnummer. 30481853
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481853 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30481853 Leverandør Toronto Research Chemicals Leverandørnr. TRCN394550100MG
 Begrænset vare
Forventet forsendelse: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Nicergoline
CAS 27848-84-6
Molekylær formel C24 H26 Br N3 O3
Mængde 100 mg
Synonym Nicergoline, Ergoline-8-methanol','10-methoxy-1,6-dimethyl-','8-(5-bromo-3-pyridinecarboxylate)','(8β)-, Ergoline-8-methanol','10-methoxy-1,6-dimethyl-','5-bromo-3-pyridinecarboxylate (ester)','(8β)- (9CI), Ergoline-8β-methanol','10-methoxy-1,6-dimethyl-','5-bromonicotinate (ester) (8CI), Indolo[4,3-fg]quinoline','ergoline-8-methanol deriv., Nicotinic acid','5-bromo-','(10-methoxy-1,6-dimethylergolin-8β-yl)methyl ester (8CI), 1,6-Dimethyl-8β-(5-bromonicotinoyloxymethyl)-10α-methoxyergoline, Cergodum, Circo-Maren, Dilasenil, Duracebrol, Ergobel, Ergotop, FI 6714, MNE, Memoq, NSC 150531, Nicergolent, Nicotergoline, Sermion, TA 079, Vasospan
InChI formel InChI=1S/C24H26BrN3O3/c1-27-13-17-8-21-24(30-3,19-5-4-6-20(27)22(17)19)9-15(12-28(21)2)14-31-23(29)16-7-18(25)11-26-10-16/h4-7,10-11,13,15,21H,8-9,12,14H2,1-3H3/t15-,21-,24+/m1/s1
SMIL CO[C@]12C[C@@H](COC(=O)c3cncc(Br)c3)CN(C)[C@@H]1Cc4cn(C)c5cccc2c45
IUPAC navn [(6aR,9R,10aS)-10a-methoxy-4,7-dimethyl-6a,8,9,10-tetrahydro-6H-indolo[4,3-fg]kinolin-9-yl]methyl 5-bromopyridin-3-carboxylat.
Molekylvægt (g/mol) 484.39
Formel vægt 483.1158
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.