Få mere at vide
Nicergoline EP Impurity C, Mikromol™
Beskrivelse
Nicergoline EP Impurity C, Mikromol
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methylmethoxylumilysergol |
| CAS | 35155-28-3 |
| Analyt- eller komponentnavne | Methylmethoxylumilysergol |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H24 N2 O2 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | [(6aR,9R,10aS)-10a-Methoxy-4,7-dimethyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinolin-9-yl]methanol (Methylmethoxylumilysergol), [(6aR,9R,10aS)-10a-Methoxy-4,7-dimethyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinolin-9-yl]methanol, Nicergoline Imp. C (EP), Methylmethoxylumilysergol, Ergoline-8-methanol, 10-methoxy-1,6-dimethyl-, (8beta)-, Lumilysergol, 10O,1-dimethyl- (7CI), (8beta)-10-Methoxy-1,6-dimethylergoline-8-methanol, Indolo[4,3-fg]quinoline, ergoline-8-methanol deriv., (+)-(5R,10S)-Meluol, 1,6-Dimethyl-8beta-(hydroxymethyl)-10alpha-methoxyergoline, 1-Methyl-10alpha-methoxydihydrolysergol, 10-Methoxy-1,6-dimethylergoline-8beta-methanol, 10alpha-Methoxy-1-methyl-9,10-dihydrolysergol, 10alpha-Methoxy-1-methyldihydrolysergol, O10,1-Dimethyllumilysergol, Nicergoline Impurity C |
| InChI formel | InChI=1S/C18H24N2O2/c1-19-10-13-7-16-18(22-3,8-12(11-21)9-20(16)2)14-5-4-6-15(19)17(13)14/h4-6,10,12,16,21H,7-9,11H2,1-3H3/t12-,16-,18+/m1/s1 |
| Løsningstype | Neat |
| SMIL | CO[C@]12C[C@@H](CO)CN(C)[C@@H]1Cc3cn(C)c4cccc2c34 |
| Vis mere |