Få mere at vide
Tocris Bioscience™ NICE 01
Mængde:
5 mg
Beskrivelse
NICE 01 is a bifunctional molecule consisting of (+)-JQ1 (Cat. No. 4499) and AP 1867 (Cat. No. 6207), with a PEG2-diamine linker. It has a high affinity (Kd = 94 pM) for FKBPF36V tagged proteins. It can induce nuclear localization of cytosolic proteins and target transcriptional regulation. It can be used to calculate the kinetic constant and passive diffusion across the nuclear pore.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (R)-1-(3-((14-((S)-4-(4-Chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)-2,13-dioxo-6,9-dioxa-3,12-diazatetradecyl)oxy)phenyl)-3-(3,4-dimethoxyphenyl)propyl (S)-1-((S)-2-(3,4,5-trimethoxyphenyl)butanoyl)piperidine-2- |
| Mængde | 5 mg |
| Mål | Protein Dimerizers |
| Molekylær formel | C63H76ClN7O13S |
| Renhed | 98% |