Få mere at vide
Nicametate, TRC
Artikelnummer. 30413279
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30413279 500 mg
30408865 50 mg
30416243 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30413279 Leverandør Toronto Research Chemicals Leverandørnr. TRCN900825500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3099-52-3
Molekylær formel C12 H18 N2 O2
Molekylvægt (g/mol) 222.28
Synonym Nicotinic Acid 2-Diethylamino-Ethyl Ester, 3-Pyridinecarboxylic acid, 2-(diethylamino)ethyl ester, Nicotinic acid, 2-(diethylamino)ethyl ester (7CI,8CI), 2-(Diethylamino)ethyl 3-pyridinecarboxylate, 2-(Diethylamino)ethyl nicotinate, Nicametate, beta-Diethylaminoethyl nicotinate
IUPAC navn 2-(diethylamino)ethyl pyridine-3-carboxylate
SMIL CCN(CC)CCOC(=O)c1cccnc1
Tekniske data
Kemisk navn eller materiale Nicametate
Renhedsgrad noter HPLC
Mængde 500 mg
InChI formel InChI=1S/C12H18N2O2/c1-3-14(4-2)8-9-16-12(15)11-6-5-7-13-10-11/h5-7,10H,3-4,8-9H2,1-2H3
Formel vægt 222.1368
Procent renhed >95%