Få mere at vide
Invitrogen™ Niban-like protein Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116934
Beskrivelse
Positive Samples: A375, K-562 Immunogen sequence: TFETLLHQEL GKGPTKEELC KSIQRVLERV LKKYDYDSSS VRKRFFREAL LQISIPFLLK KLAPTCKSEL PRFQELIFED FARFILVENT YEEVVLQTVM KDILQAVKEA AVQRKHNLYR DSMVMHNSDP NLHLLAEGAP IDWGEEYSNS GGGGSPSPST PESATLSEKR RRAKQVVSVV QDEEVGLPFE ASPESPPPAS PDGVTEIRGL LAQGLRPESP PPAGPLLNGA PAGESPQPKA APEASSPPAS PLQHLLPGKA VDLGPPKPSD QETGEQVSSP SSHPALHTTT EDSAGVQTEF.
FAM129B (Family With Sequence Similarity 129 Member B, Niban Like Protein 1) is a Protein Coding gene. May play a role in apoptosis suppression. May promote melanoma cell invasion in vitro. Among its related pathways are MAP Kinase Signaling. An important paralog of this gene is FAM129A.
Tekniske data
| Niban-like protein | |
| Polyclonal | |
| Unconjugated | |
| Fam129b | |
| 9130404D14Rik; bA356B19.6; C9orf88; FAM129B; family with sequence similarity 129 member B; family with sequence similarity 129, member B; Meg-3; Melanoma invasion by ERK; MINERVA; NIBAN2; Niban-like protein 1; OC58; Protein FAM129B; Protein Niban 2; RGD1307018 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64855 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.32 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q96TA1 | |
| Fam129b | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 447-746 of human FAM129B (NP_0737442). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.