missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Niban-like protein Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5148293
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Niban-like protein Polyclonal Antibody for IHC (P)
FAM129B (Family With Sequence Similarity 129 Member B, Niban Like Protein 1) is a Protein Coding gene. May play a role in apoptosis suppression. May promote melanoma cell invasion in vitro. Among its related pathways are MAP Kinase Signaling. An important paralog of this gene is FAM129A.
Tekniske data
| Niban-like protein | |
| Polyclonal | |
| Unconjugated | |
| Fam129b | |
| 9130404D14Rik; bA356B19.6; C9orf88; FAM129B; family with sequence similarity 129 member B; family with sequence similarity 129, member B; Meg-3; Melanoma invasion by ERK; MINERVA; NIBAN2; Niban-like protein 1; OC58; Protein FAM129B; Protein Niban 2; RGD1307018 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 64855 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 100 μg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q96TA1 | |
| Fam129b | |
| E. coli-derived Human FAM129B fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion