Få mere at vide
Niacinamide Ascorbate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Nicotinamide Ascorbate |
| CAS | 1987-71-9 |
| Molekylær formel | C6 H8 O6 . C6 H6 N2 O |
| Mængde | 100 mg |
| Synonym | Niacinamide Ascorbate, L-Ascorbic acid','mixt. with 3-pyridinecarboxamide, L-Ascorbic acid','mixt. with nicotinamide (8CI), 3-Pyridinecarboxamide','mixt. contg. (9CI), Ascorbic acid nicotinamide complex, Ascorbic acid-niacinamide complex, KA 1015, Merpress, NSC 13130, Niacinamide ascorbate, Nicastubin, Nicastubine, Nicoscorbine, Nicotinamide ascorbate, Nicotinamide','L-ascorbate, Vitamin C-nicotinamide mixt. |
| InChI formel | InChI=1S/C6H6N2O.C6H8O6/c7-6(9)5-2-1-3-8-4-5;7-1-2(8)5-3(9)4(10)6(11)12-5/h1-4H,(H2,7,9);2,5,7-10H,1H2/t;2-,5+/m.0/s1 |
| SMIL | NC(=O)c1cccnc1.OC[C@H](O)[C@H]2OC(=O)C(=C2O)O |
| IUPAC navn | (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one;pyridine-3-carboxamide |
| Molekylvægt (g/mol) | 298.249 |
| Formel vægt | 298.08 |