missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
NHA2/SLC9B2/NHEDC2 Antibody (896151), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8404MFV500
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
NHA2/SLC9B2/NHEDC2 Monoclonal antibody specifically detects NHA2/SLC9B2/NHEDC2 in Human samples. It is validated for Flow Cytometry, CyTOF
Tekniske data
| NHA2/SLC9B2/NHEDC2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 133308 | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 896151 | |
| Flow Cytometry, CyTOF-ready | |
| FLJ23984, Mitochondrial Na(+)/H(+) exchanger NHA2, mitochondrial sodium/hydrogen exchanger NHA2, Na(+)/H(+) exchanger-like domain-containing protein 2, Na+/H+ exchanger domain containing 2, NHA2NHE10, NHE domain-containing protein 2, Sodium/hydrogen exchanger-like domain-containing protein 2 | |
| NS0 mouse myeloma cell line transfected with human NHA2/SLC9B2, Met1-Val537, Accession # NP_849155 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion