missing translation for 'onlineSavingsMsg'
Få mere at vide

NG,NG-Dimethylarginine-d6 Dihydrochloride, TRC

Artikelnummer. 30377609
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30377609 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30377609 Leverandør Toronto Research Chemicals Leverandørnr. TRCD46357725MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale NG,NG-Dimethylarginine-D6 Dihydrochloride
CAS 1313730-20-9
Renhedsgrad noter HPLC
Molekylær formel C8H14D6Cl2N4O2
Mængde 25 mg
Synonym (2S)-2-Amino-5-[(N,N-bis(trideuteriomethyl)carbamimidoyl)amino]pentanoic acid dihydrochloride, NG,NG-Dimethylarginine-d6 dihydrochloride, NG,NG-Dimethyl-L-arginine-D6 (NG,NG-dimethyl-D6) dihydrochloride, NG,NG-Di(methyl-d3)-L-arginine dihydrochloride, N5-[[Di(methyl-d3)amino]iminomethyl]-L-ornithine dihydrochloride
InChI formel InChI=1S/C8H18N4O2/c1-12(2)8(10)11-5-3-4-6(9)7(13)14/h6H,3-5,9H2,1-2H3,(H2,10,11)(H,13,14)/t6-/m0/s1
SMIL Cl.Cl.[2H]C([2H])([2H])N(C(=N)NCCC[C@H](N)C(=O)O)C([2H])([2H])[2H]
Molekylvægt (g/mol) 281.21
Formel vægt 280.134
Procent renhed >95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.