Få mere at vide
NG,NG’-Dimethyl-L-arginine-D6, TRC
Artikelnummer. 30375429
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30375429 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30375429 Leverandør Toronto Research Chemicals Leverandørnr. TRCD46358210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale NG,NG'-Dimethyl-L-arginine-d6
CAS 1331888-08-4
Renhedsgrad noter HPLC
Molekylær formel C8H12D6N4O2
Mængde 10 mg
Synonym N5-[(Dimethylamino)iminomethyl]-L-ornithine-d6, N5-[Bis(methylamino)methylene]-L-ornithine-d6, SDMA-d6
InChI formel InChI=1S/C8H18N4O2/c1-10-8(11-2)12-5-3-4-6(9)7(13)14/h6H,3-5,9H2,1-2H3,(H,13,14)(H2,10,11,12)/t6-/m0/s1/i1D3,2D3
SMIL N[C@H](C(O)=O)CCC/N=C(NC([2H])([2H])[2H])/NC([2H])([2H])[2H]
Molekylvægt (g/mol) 208.29
Formel vægt 208.1806
Procent renhed >95