missing translation for 'onlineSavingsMsg'
Få mere at vide

NG-monomethyl-L-argininacetat, TRC

Artikelnummer. 30474076
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30474076 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30474076 Leverandør Toronto Research Chemicals Leverandørnr. TRCM565000100MG
 Begrænset vare
Forventet forsendelse: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale NG-Monomethyl-L-arginine Acetate
CAS 53308-83-1
Renhedsgrad noter HPLC
Molekylær formel C7 H16 N4 O2 . C2 H4 O2
Mængde 100 mg
Synonym N5-[Imino(methylamino)methyl]-L-ornithine Acetate, L-NMMA, ANO 1020, Tilarginine Acetate
InChI formel InChI=1S/C7H16N4O2.C2H4O2/c1-10-7(9)11-4-2-3-5(8)6(12)13;1-2(3)4/h5H,2-4,8H2,1H3,(H,12,13)(H3,9,10,11);1H3,(H,3,4)/t5-;/m0./s1
SMIL CNC(=N)NCCC[C@H](N)C(=O)O.CC(=O)O
IUPAC navn eddikesyre; (2S)-2-amino-5-[(N-methylcarbamimidoyl)amino]pentansyre
Molekylvægt (g/mol) 248.28
Formel vægt 248.1485
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.