Få mere at vide
Tocris Bioscience™ NFPS
GREENER_CHOICE
Artikelnummer. 18004172
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18004172 10 mg
18025662 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18004172 Leverandør Tocris Bioscience™ Leverandørnr. 2789/10
Selective non-transportable GlyT1 inhibitor
Selective, non-transportable inhibitor of GlyT1 which displays no activity at GABA or glutamate receptors (IC50 values are 2.8, 9.8 and 56000 nM for hGlyT1, rGlyT1 and rGlyT2 respectively). Enhances the amplitude of the NMDA component of glutamatergic EPSCs.
R-enantiomer also available.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 405225-21-0
Molekylær formel C24H24FNO3
Tekniske data
Kemisk navn eller materiale NFPS
Mængde 10 mg
Formel vægt 393.45
Renhed >98%
Molekylvægt (g/mol) 393.458