missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFIA Antibody (PCRP-NFIA-2C6), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14016JF646
Additional Details : Weight : 0.00970kg
Description
NFIA Monoclonal antibody specifically detects NFIA in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
NFIA | |
Monoclonal | |
Janelia Fluor 646 | |
CCAAT-box-binding transcription factor, CTF, DKFZp686J23256, FLJ39164, KIAA1439DKFZp434L0422, NF1-A, NF-I/A, NFI-A, NFI-L, nuclear factor 1 A-type, Nuclear factor 1/A, nuclear factor I/ATGGCA-binding protein | |
Recombinant fragment of human NFIA protein corresponding to protein domain (Uniprot: Q15784) | |
0.1 mL | |
Cell Cycle and Replication | |
4774 | |
Store at 4°C in the dark. | |
IgG2c κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
PCRP-NFIA-2C6 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |