missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ NFAT inhibitor, Cell Permeable
GREENER_CHOICE

Artikelnummer. 18706509
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18706509 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18706509 Leverandør Tocris Bioscience™ Leverandørnr. 5710/1

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Cell permeable NFAT inhibitor

NFAT inhibitor, Cell Permeable is an NFAT inhibitor. Inhibits LPS or LPS plus IFN-γ-induced IL-12 p40, IL-12 p70, IL-23 and TNF secretion from bone marrow-derived macrophages (BMDMs). Also attenuates NO production and Nos2 mRNA expression in LPS-stimulated BMDMs. Reduces inflammation and epithelial damage in a mouse model of colitis. In an animal model of Alzheimer's disease NFAT inhibitor improves cognitive and synaptic function, reduces glial activation and lowers amyloid levels; in vitro it reduces glutamate dysregulation and neuron hyperactivity.
TRUSTED_SUSTAINABILITY

Tekniske data

CAS 592517-80-1
Indhold og opbevaring Store at -20°C
Oversigt Protein Ser/Thr Phosphatase Inhibitors
Molekylvægt (g/mol) Observed M.W.: 3573.15
Mængde 1 mg
Strukturel form C147H259N67O36S
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.