Få mere at vide
Tocris Bioscience™ NFAT inhibitor, Cell Permeable
GREENER_CHOICE
Cell permeable NFAT inhibitor
Brand:  Tocris Bioscience™ 5710/1
Encompass
Produktkode 15767066
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
NFAT inhibitor, Cell Permeable is a NFAT inhibitor. Inhibits LPS or LPS plus IFN-γ-induced IL-12 p40, IL-12 p70, IL-23 and TNF secretion from bone marrow-derived macrophages (BMDMs). Also attenuates NO production and Nos2 mRNA expression in LPS-stimulated BMDMs. Improves symptoms in a mouse model of colitis. Exhibits immunosuppressive effects; enhances graft survival in mice. Cell permeable.
Tekniske data
Tekniske data
592517-80-1
Protein Ser/Thr Phosphatase Inhibitors
1 mg
Store at -20°C
Observed M.W.: 3573.15
C147H259N67O36S
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner